7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 7-tert-Butyl 2-ethyl 5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate

A198382-100MG A198382-1G A198382-250MG
76785-202EA 17 USD
76785-202 76785-204 76785-206
7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate 97%
7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₃O₄
MW: 295.34 g/mol
MDL Number: MFCD10566312
CAS Number: 1053656-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR