Ethyl-5-chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylate

A197780-250MG A197780-100MG
76785-186EA 95.49 USD
76785-186 77667-644
Ethyl-5-chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylate 97%
Ethyl-5-chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
MDL Number: MFCD09878687
CAS Number: 800401-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR