5-Phenylisoxazole-3-carboxamide 95%

Supplier: Ambeed
Warning

A196867-100MG A196867-250MG
76785-146EA 20.16 USD
76785-146 76785-148
5-Phenylisoxazole-3-carboxamide 95%
5-Phenylisoxazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
MDL Number: MFCD00464227
CAS Number: 23088-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR