2-((tert-Butoxycarbonyl)amino)-3-ethoxy-3-oxopropanoic acid 95%

Supplier: Ambeed
Warning

A196608-250MG A196608-25G A196608-5G A196608-1G
76785-140EA 10.61 USD
76785-140 76785-142 76785-144 77593-274
2-((tert-Butoxycarbonyl)amino)-3-ethoxy-3-oxopropanoic acid 95%
2-((tert-Butoxycarbonyl)amino)-3-ethoxy-3-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₆
MW: 247.2451 g/mol
MDL Number: MFCD09835539
CAS Number: 137401-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR