1-(tert-Butoxycarbonyl)-1,4-diazepane-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A196050-100MG A196050-50MG
76785-096EA 159.14 USD
76785-096 76785-098
1-(tert-Butoxycarbonyl)-1,4-diazepane-5-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-1,4-diazepane-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.28 g/mol
MDL Number: MFCD09752214
CAS Number: 1214824-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR