2,3-Difluoroisonicotinic acid 96%

Supplier: Ambeed
Warning

A195974-1G A195974-250MG A195974-25G A195974-5G
76785-084EA 14 USD
76785-084 76785-086 76785-088 76785-090
2,3-Difluoroisonicotinic acid 96%
2,3-Difluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MDL Number: MFCD09031174
CAS Number: 851386-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR