1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester

A195241-100MG A195241-250MG A195241-5G A195241-1G
76785-078EA 15.92 USD
76785-078 76785-080 76785-082 77593-260
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₂
MW: 273.18 g/mol
MDL Number: MFCD08060504
CAS Number: 852227-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR