4-Bromo-5-fluoro-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

A194682-100MG A194682-1G
76785-068EA 26 USD
76785-068 76785-070
4-Bromo-5-fluoro-2-hydroxypyridine 98%
4-Bromo-5-fluoro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MDL Number: MFCD05662410
CAS Number: 884495-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR