Vardenafil hydrochloride trihydrate 98%

Supplier: Ambeed
Warning

A194638-10MG A194638-1G A194638-250MG A194638-25MG A194638-50MG A194638-5G A194638-1MG A194638-5MG A194638-100MG A194638-25G
76785-056EA 10 USD
76785-056 76785-058 76785-060 76785-062 76785-064 76785-066 77600-578 77600-580 77871-968 77871-969
Vardenafil hydrochloride trihydrate 98%
Vardenafil hydrochloride trihydrate

Precaución: for Research Use Only

Formula: C₂₃H₃₉ClN₆O₇S
MDL Number: MFCD08458876
CAS Number: 330808-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR