Methyl-4-nitro-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A193303-100G A193303-10G A193303-1G A193303-250MG A193303-25G A193303-5G
76785-002EA 235 USD
76785-002 76785-004 76785-006 76785-008 76785-010 76785-012
Methyl-4-nitro-1H-pyrazole-3-carboxylate 97%
Methyl-4-nitro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅N₃O₄
MDL Number: MFCD00505604
CAS Number: 138786-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR