Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-fluorophenyl)butanoic acid

A192863-1G A192863-250MG A192863-5G
76784-996EA 160.2 USD
76784-996 76784-998 77600-560
Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid 97%
Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀FNO₄
MW: 297.33 g/mol
MDL Number: MFCD01076280
CAS Number: 218609-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR