3-Bromopyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A191864-100MG A191864-1G A191864-5G A191864-250MG A191864-25G
76784-968EA 24 USD
76784-968 76784-970 76784-972 77871-957 77871-958
3-Bromopyrazine-2-carboxylic acid 97%
3-Bromopyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C5H3BrN2O2
MDL Number: MFCD08753934
CAS Number: 937669-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR