Methyl-2,3-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,3-dihydroxybenzoate

A191618-10G A191618-1G A191618-5G
76784-962EA 31.83 USD
76784-962 76784-964 76784-966
Methyl-2,3-dihydroxybenzoate 98%
Methyl 2,3-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.1467 g/mol
MDL Number: MFCD00039520
CAS Number: 2411-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR