Methyl-4-bromo-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1H-indole-2-carboxylate

A191445-100MG A191445-1G A191445-250MG A191445-25G A191445-5G
76784-952EA 16.98 USD
76784-952 76784-954 76784-956 76784-958 76784-960
Methyl-4-bromo-1H-indole-2-carboxylate 98%
Methyl-4-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.09 g/mol
MDL Number: MFCD04966956
CAS Number: 167479-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR