5-Bromo-1,2,3-trichlorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4,5-trichlorobenzene

A191442-100G A191442-10G A191442-1G A191442-25G A191442-500G A191442-5G
76784-940EA 90 USD
76784-940 76784-942 76784-944 76784-946 76784-948 76784-950
5-Bromo-1,2,3-trichlorobenzene 98%
5-Bromo-1,2,3-trichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₃
MDL Number: MFCD00155009
CAS Number: 21928-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR