Nilotinib monohydrochloride monohydrate 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-N-(3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl)-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzamide hydrochloride hydrate

A191440-100MG A191440-50MG A191440-5G A191440-1G
76784-934EA 7.43 USD
76784-934 76784-936 76784-938 77667-598
Nilotinib monohydrochloride monohydrate 98+%
Nilotinib monohydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₂₈H₂₅ClF₃N₇O₂
MW: 583.99 g/mol
MDL Number: MFCD18251359
CAS Number: 923288-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR