Tetrahydroxy-1,4-benzoquinone disodium salt 95%

Supplier: Ambeed
Warning

Synonyms: Sodium 4,5-Dihydroxy-3,6-dioxocyclohexa-1,4-diene-1,2-bis(olate), Tetrahydroxyquinone Disodium Salt

A191134-100G A191134-1G A191134-25G A191134-5G
76784-920EA 432 USD
76784-920 76784-922 76784-924 76784-926 76784-928
Tetrahydroxy-1,4-benzoquinone disodium salt 95%
Tetrahydroxy-1,4-benzoquinone disodium salt

Precaución: for Research Use Only

Formula: C₆H₄O₆
MW: 216.06 g/mol
MDL Number: MFCD00036127
CAS Number: 1887-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR