8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A190896-100MG A190896-1G A190896-250MG A190896-5G
76784-908EA 26.53 USD
76784-908 76784-910 76784-912 76784-914
8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MW: 248.5500 g/mol
MDL Number: MFCD08752231
CAS Number: 1159813-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR