7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%
Supplier: Ambeed
|
Warning
|
A190679-100MG
A190679-1G
A190679-250MG
76784-892EA
51
USD
76784-892
76784-894
76784-896
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃O |
MDL Number:
MFCD09999206 CAS Number: 93587-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...