(±)-3,7-Dimethyl-3-octanol 97%

Supplier: Ambeed
Warning

Synonyms: Tetrahydrolinalool, 3,7-Dimethyloctan-3-ol, 3,7-Dimethyl-3-octanol

A190618-100G A190618-10G A190618-500G A190618-25G
76784-886EA 15.92 USD
76784-886 76784-888 76784-890 77084-720
(±)-3,7-Dimethyl-3-octanol 97%
(±)-3,7-Dimethyl-3-octanol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
MDL Number: MFCD00004482
CAS Number: 78-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR