5-Methoxypyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A189950-100G A189950-1G A189950-25G A189950-5G
76784-876EA 223 USD
76784-876 76784-878 76784-880 76784-882
5-Methoxypyrazine-2-carboxylic acid 97%
5-Methoxypyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C6H6N2O3
MDL Number: MFCD10697773
CAS Number: 40155-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR