(5-Bromo-2-chlorophenyl)methanamine 95%

Supplier: Ambeed
Warning

A189023-250MG A189023-10G
76784-860EA 28.65 USD
76784-860 77667-574
(5-Bromo-2-chlorophenyl)methanamine 95%
(5-Bromo-2-chlorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.4900 g/mol
MDL Number: MFCD12026286
CAS Number: 1096296-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR