2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
Supplier: Ambeed
|
Warning
|
A188862-100MG
A188862-1G
A188862-5G
76784-856EA
6
USD
76784-856
76784-858
77871-946
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
2'-Methoxy-5'-(trifluoromethyl)acetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O₂ |
MDL Number:
MFCD09832300 CAS Number: 503464-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...