trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

A188347-100G A188347-1G A188347-25G A188347-5G
76784-846EA 127 USD
76784-846 76784-848 76784-850 76784-852
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl 97%
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₁H₂₅F
MDL Number: MFCD11053438
CAS Number: 87260-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR