exo-Tetrahydrodicyclopentadiene 94%

Supplier: Ambeed
Danger

Synonyms: exo-Tricyclo[5.2.1.0(2,6)]decane, (3aR,4S,7R,7aS)-rel-Octahydro-1H-4,7-methanoindene

A188304-25G A188304-5G
76784-838EA 51.99 USD
76784-838 76784-840
exo-Tetrahydrodicyclopentadiene 94%
exo-Tetrahydrodicyclopentadiene

Precaución: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD01714931
CAS Number: 2825-82-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR