Thieno[3,2-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

A187616-1G A187616-25G A187616-5G
76784-802EA 13 USD
76784-802 76784-804 76784-806
Thieno[3,2-d]pyrimidin-4(3H)-one 98%
Thieno[3,2-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₄N₂OS
MDL Number: MFCD03407527
CAS Number: 16234-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR