(2S,3S)-2-((2S,3S)-2-(((Benzyloxy)carbonyl)amino)-3-methylpentanamido)-3-methylpentanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Z-Ile-Ile

A187528-1G A187528-250MG A187528-25G A187528-5G
76784-784EA 28.65 USD
76784-784 76784-786 76784-788 76784-790
(2S,3S)-2-((2S,3S)-2-(((Benzyloxy)carbonyl)amino)-3-methylpentanamido)-3-methylpentanoic acid 95%
(2S,3S)-2-((2S,3S)-2-(((Benzyloxy)carbonyl)amino)-3-methylpentanamido)-3-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₀N₂O₅
MW: 378.46 g/mol
MDL Number: MFCD00027066
CAS Number: 42538-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR