2-(Aminomethyl)benzonitrile 97%

Supplier: Ambeed
Danger

A187487-100MG A187487-1G A187487-250MG A187487-5G
76784-776EA 32 USD
76784-776 76784-778 76784-780 76784-782
2-(Aminomethyl)benzonitrile 97%
2-(Aminomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD06797831
CAS Number: 344957-25-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR