3-[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]benzoic acid 98%

Supplier: Ambeed
Warning

A186849-100MG A186849-1G A186849-250MG
76784-764EA 7 USD
76784-764 76784-766 76784-768
3-[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]benzoic acid 98%
3-[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₅F₃N₂O₃
MDL Number: MFCD09907879
CAS Number: 1092400-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR