6-Chloroquinoline-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A186189-100MG A186189-1G A186189-250MG
76784-732EA 41 USD
76784-732 76784-734 76784-736
6-Chloroquinoline-4-carboxylic acid 95%
6-Chloroquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD07506540
CAS Number: 62482-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR