Z-D-glu-OMe 98%

Supplier: Ambeed
Warning

A185886-5G A185886-100G A185886-25G
76784-730EA 58 USD
76784-730 77667-550 77871-925
Z-D-glu-OMe 98%
Z-D-glu-OMe

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₆
MDL Number: MFCD00798654
CAS Number: 26566-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR