1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A185616-100MG
A185616-1G
A185616-250MG
76784-724EA
58.35
USD
76784-724
76784-726
76784-728
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride 95%
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₄ClNO₂ MW: 191.6600 g/mol |
MDL Number:
MFCD11869757 CAS Number: 374794-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...