Tetrabutylammonium thiocyanate 97%

Supplier: Ambeed
Warning

Synonyms: Tetrabutylammonium Rhodanide

A185554-100G A185554-10G A185554-25G A185554-5G
76784-716EA 391 USD
76784-716 76784-718 76784-720 76784-722
Tetrabutylammonium thiocyanate 97%
Tetrabutylammonium thiocyanate

Precaución: for Research Use Only

Formula: C₁₇H₃₆N₂S
MW: 300.55 g/mol
MDL Number: MFCD00012140
CAS Number: 3674-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR