Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphite, Phosphorous acid tris(1,1,1,3,3,3-hexafluoro-2-propyl) ester
A184506-250MG
A184506-1G
76784-704EA
24
USD
76784-704
77871-918
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite 98%
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Precaución: for Research Use Only
Formula:
C₉H₃F₁₈O₃P |
MDL Number:
MFCD00192561 CAS Number: 66470-81-3 UN: 1993 ADR: 3,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...