Methyl-3-amino-5-chlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-chlorobenzoate

A184447-1G A184447-250MG A184447-5G
76784-698EA 45.62 USD
76784-698 76784-700 76784-702
Methyl-3-amino-5-chlorobenzoate 98%
Methyl 3-amino-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD11053764
CAS Number: 21961-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR