(S)-1-N-Cbz-Pipecolinic acid 98%

Supplier: Ambeed
Warning

A183829-10G A183829-250MG A183829-5G A183829-1G
76784-678EA 87 USD
76784-678 76784-680 76784-682 77593-180
(S)-1-N-Cbz-Pipecolinic acid 98%
(S)-1-N-Cbz-Pipecolinic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.29 g/mol
MDL Number: MFCD01863645
CAS Number: 28697-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR