1-(2-Amino-5-chlorophenyl)ethanone 97%

Supplier: Ambeed
Warning

A183542-100MG A183542-1G A183542-250MG A183542-5G
76784-662EA 15.92 USD
76784-662 76784-664 76784-666 76784-668
1-(2-Amino-5-chlorophenyl)ethanone 97%
1-(2-Amino-5-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.608 g/mol
MDL Number: MFCD07434611
CAS Number: 1685-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR