Zinc monomethionine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-((2-Amino-4-(methylthio)butanoyl)oxy)zinc(II) hydrogensulphate

A181949-10G A181949-1G A181949-250MG A181949-25G A181949-5G
76784-608EA 27 USD
76784-608 76784-610 76784-612 76784-614 76784-616
Zinc monomethionine 95%
Zinc monomethionine

Precaución: for Research Use Only

Formula: C₅H₁₀SNO₂Zn·H₂SO₄
MDL Number: MFCD04118310
CAS Number: 56329-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR