2-Methoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol 95% contains isomer

Supplier: Ambeed
Warning

A181455-100G A181455-25G
76784-586EA 23.34 USD
76784-586 76784-588
2-Methoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol 95% contains isomer
2-Methoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.22 g/mol
MDL Number: MFCD00036666
CAS Number: 68527-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR