trans-4-Aminoadamantan-1-ol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: trans-1-Amino-4-hydroxyadamantane hydrochloride, trans-4-Amino-1-adamantanol hydrochloride

A180906-100G A180906-1G A180906-250MG A180906-25G A180906-5G
76784-574EA 356.47 USD
76784-574 76784-576 76784-578 76784-580 76784-582
trans-4-Aminoadamantan-1-ol hydrochloride 97%
trans-4-Aminoadamantan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₈ClNO
MW: 203.7100 g/mol
MDL Number: MFCD11099880
CAS Number: 62075-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR