[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-((4-Chloro-6-((2,3-dimethylphenyl)amino)pyrimidin-2-yl)thio)acetic acid, WY-14643, Pirinixic acid

A180605-10MG A180605-1G A180605-1MG A180605-250MG A180605-50MG A180605-100MG
76784-554EA 7.43 USD
76784-554 76784-556 76784-558 76784-560 76784-562 77667-494
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid 98%
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClN₃O₂S
MW: 323.80 g/mol
MDL Number: MFCD00191335
CAS Number: 50892-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR