Sulfadimethoxine 98+%

Supplier: Ambeed
Warning

A178691-1MG A178691-25MG A178691-1G A178691-100G A178691-25G A178691-5G A178691-500G
76784-516EA 5 USD
76784-516 76784-518 77567-632 77600-448 77667-478 77667-480 77871-894
Sulfadimethoxine 98+%
Sulfadimethoxine

Precaución: For research use only.

Formula: C₁₂H₁₄N₄O₄S
MDL Number: MFCD00057345
CAS Number: 122-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR