(2S,3S)-2-(Benzyl(methyl)amino)-3-methylpentanoic acid 97+%

Supplier: Ambeed
Warning

A178413-1G A178413-250MG A178413-5G
76784-502EA 47.75 USD
76784-502 76784-504 76784-506
(2S,3S)-2-(Benzyl(methyl)amino)-3-methylpentanoic acid 97+%
(2S,3S)-2-(Benzyl(methyl)amino)-3-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₂
MW: 235.32 g/mol
MDL Number: MFCD01632094
CAS Number: 4125-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR