tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate, 3-(Hydroxymethyl)-2-methyl-1H-indole, N-BOC protected
A178159-10G
A178159-1G
A178159-250MG
A178159-5G
76784-488EA
126.25
USD
76784-488
76784-490
76784-492
76784-494
tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₃ MW: 261.3163 g/mol |
MDL Number:
MFCD05864723 CAS Number: 914349-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...