Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-3-yl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(6-bromoimidazo[1,2-a]pyridin-3-yl)acetate

A177235-100MG A177235-1G A177235-250MG A177235-5G
76784-476EA 36.08 USD
76784-476 76784-478 76784-480 76784-482
Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-3-yl)acetate 95%
Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
MDL Number: MFCD09839780
CAS Number: 603311-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR