2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A176178-100MG A176178-1G A176178-250MG A176178-5G
76784-444EA 55.17 USD
76784-444 76784-446 76784-448 76784-450
2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid 95%
2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₅Cl₂NO₂S
MDL Number: MFCD07376343
CAS Number: 1178420-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR