tert-Butyl 4-oxocyclohexane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-oxocyclohexanecarboxylate, 4-(tert-Butoxycarbonyl)cyclohexan-1-one, 1-(tert-Butoxycarbonyl)-4-oxocyclohexane

A175881-1G A175881-250MG A175881-5G
76784-434EA 98.67 USD
76784-434 76784-436 76784-438
tert-Butyl 4-oxocyclohexane-1-carboxylate 97%
tert-Butyl 4-oxocyclohexane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₃
MW: 198.259 g/mol
MDL Number: MFCD08061192
CAS Number: 38446-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR