ZM-336372 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Dimethylamino)-N-(3-(4-hydroxybenzamido)-4-methylphenyl)benzamide, ZM 336372

A175274-100MG A175274-10MG A175274-1MG A175274-50MG A175274-5MG
76784-400EA 163.38 USD
76784-400 76784-402 76784-404 76784-406 76784-408
ZM-336372 98%
ZM-336372

Precaución: for Research Use Only

Formula: C₂₃H₂₃N₃O₃
MW: 389.455 g/mol
MDL Number: MFCD02683971
CAS Number: 208260-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR