trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl, trans,trans-4-(3,4-Difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)

A174976-10G A174976-1G
76784-388EA 23 USD
76784-388 76784-390
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl

Precaución: for Research Use Only

Formula: C₂₁H₃₀F₂
MW: 320.468 g/mol
MDL Number: MFCD09838997
CAS Number: 82832-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR