4-Bromo-2-methylisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A174795-10G A174795-250MG A174795-25G A174795-5G A174795-1G
76784-380EA 267 USD
76784-380 76784-382 76784-384 76784-386 77871-874
4-Bromo-2-methylisoquinolin-1(2H)-one 97%
4-Bromo-2-methylisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD00489133
CAS Number: 33930-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR